Learn More
MilliporeSigma™ Supelco™ Eppendorf™ CH-500 HPLC column Heater System
Catalog No. 111005796 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
UK cord
Catalog No. Description
11-100-5796 UK cord
1 options
Catalog No. 11-100-5796 Supplier MilliporeSigma™ Supelco™ Supplier No. 500860
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This heater/controller essentially combines the features of the CH-30 heater and the TC-50 controller in a single unit. Itincludes Eppendorf innovative reversible heating blocks and remote heater on/off capability. Alternatively, the heatingchannels for 1/8 in. columns can be used to heat solvent before it enters the injector.CE/CSA/UL approved.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC
Description UK cord
Eppendorf is a registered trademark of Eppendorf AG