Learn More
MilliporeSigma™ Supelco™ Eppendorf™ CH-500 HPLC column Heater System
Catalog No. p-7194181 Shop All MilliporeSigma Supelco Products
Click to view available options
Description:
UK cord
This heater/controller essentially combines the features of the CH-30 heater and the TC-50 controller in a single unit. Itincludes Eppendorf innovative reversible heating blocks and remote heater on/off capability. Alternatively, the heatingchannels for 1/8 in. columns can be used to heat solvent before it enters the injector.CE/CSA/UL approved.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC
Description UK cord
Eppendorf is a registered trademark of Eppendorf AG
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.