Learn More
Electron Microscopy Sciences Eppendorf Chuck; RMC
SDP
Catalog No. 5029556
Encompass_Preferred
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes. Which have been polymerized in Eppendorf tubes.
Catalog No. 50-295-56 Supplier Electron Microscopy Sciences Supplier No. 69992
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes. Which have been polymerized in Eppendorf tubes.