Learn More
Eppendorf 3P Eppendorf DNA LoBind Microcentrifuge Tubes, 1.5 mL, PCR Clean, 250/Pk.
Catalog No. NC3920193
Encompass_Preferred
These 1.5 mL snap cap microtubes are engineered from high-quality polypropylene to minimize sample-to-surface binding, ensuring maximal recovery of DNA and RNA molecules without the need for additional surface coatings. Designed for secure sample preparation and storage, these tubes are suitable for forensic analysis, microarrays, and Next Generation Sequencing applications.
  • Provides near 100 % recovery of nucleic acid molecules.
  • Certified PCR clean, ensuring freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Features precise lid sealing to minimize evaporation rates.
  • Centrifugation stability rated up to 30,000 ×g.
  • Manufactured without slip agents, plasticizers, or biocides.
  • Operational temperature range of -86 to 100 °C.
  • Autoclavable when opened at 121 °C for 20 minutes.
  • Supplied in a pack of 250 tubes across 5 bags.
Catalog No. NC3920193 Supplier Eppendorf 3P Supplier No. 022431021
These 1.5 mL snap cap microtubes are engineered from high-quality polypropylene to minimize sample-to-surface binding, ensuring maximal recovery of DNA and RNA molecules without the need for additional surface coatings. Designed for secure sample preparation and storage, these tubes are suitable for forensic analysis, microarrays, and Next Generation Sequencing applications.
  • Provides near 100 % recovery of nucleic acid molecules.
  • Certified PCR clean, ensuring freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Features precise lid sealing to minimize evaporation rates.
  • Centrifugation stability rated up to 30,000 ×g.
  • Manufactured without slip agents, plasticizers, or biocides.
  • Operational temperature range of -86 to 100 °C.
  • Autoclavable when opened at 121 °C for 20 minutes.
  • Supplied in a pack of 250 tubes across 5 bags.