Learn More
Pipette Supplies, Inc. EPPENDORF PIVOT GREASE 20ML
SDP
Catalog No. NC1330424
Encompass
Eppendorf 20mL pivot grease (also called Peg Fat) for rotor to lubricate the pivot joints on swing-bucket rotors.Eppendorf International Part # 5810 350.050, 5810350010Made in Germany
Catalog No. NC1330424 Supplier Pipette Supplies, Inc. Supplier No. EP 022634330
Eppendorf 20mL pivot grease (also called Peg Fat) for rotor to lubricate the pivot joints on swing-bucket rotors.Eppendorf International Part # 5810 350.050, 5810350010Made in Germany