Learn More
Pipette Supplies, Inc. Eppendorf Power Supply with Power Plug Adapters / Pipette Charger (Eppendorf)
SDP
Catalog No. NC3071610
Encompass
Eppendorf Power Supply with Power Plug Adapters / Pipette Charger.Power supply (240V50-60Hz) for the Charger Stands including EP 3116000040, and also for the following pipettes, multi-dispensers and pipette controllers:Easypet 3Repeater E3, E3x / Multipette E3, E3xRepeater Plus / Multipette Plusstream / XstreamXplorer / Xplorer PlusNote: Wall to pipette charging.100 – 240 VAC / 5 VDCSame as part # EP 022462423Not compatible with the older Easypet. The Easypet power supply is EP 022233602.
Catalog No. NC3071610 Supplier Pipette Supplies, Inc. Supplier No. EP 4986603005
Eppendorf Power Supply with Power Plug Adapters / Pipette Charger.Power supply (240V50-60Hz) for the Charger Stands including EP 3116000040, and also for the following pipettes, multi-dispensers and pipette controllers:Easypet 3Repeater E3, E3x / Multipette E3, E3xRepeater Plus / Multipette Plusstream / XstreamXplorer / Xplorer PlusNote: Wall to pipette charging.100 – 240 VAC / 5 VDCSame as part # EP 022462423Not compatible with the older Easypet. The Easypet power supply is EP 022233602.