Learn More
Next Advance Inc Navy Eppendorf RNA Lysis Kit 50 pack (1.5 mL)
SDP
Catalog No. NC1785068
Encompass
Bead lysis kits for the homogenization of large, tough samples. Pack of 50, 1.5 mL Eppendorf Safe-Lock tubes prefilled with stainless steel beads. RNase free.
Catalog No. NC1785068 Supplier Next Advance Inc Supplier No. NAVYE1RNA
Bead lysis kits for the homogenization of large, tough samples. Pack of 50, 1.5 mL Eppendorf Safe-Lock tubes prefilled with stainless steel beads. RNase free.