Learn More
USA Scientific Inc Eppendorf Tube 5.0 mL LoBind
Catalog No. 502745322
Encompass
Eppendorf Protein LoBind Tube 5.0 mL. Conical bottom polypropylene centrifuge tube with flip-open cap. 2 bags of 50 tubes (100).
Catalog No. 50-274-5322 Supplier USA Scientific Inc Supplier No. 40118302
Eppendorf Protein LoBind Tube 5.0 mL. Conical bottom polypropylene centrifuge tube with flip-open cap. 2 bags of 50 tubes (100).