Learn More
USA Scientific Inc Eppendorf Tubes 5 mL screw cap
Supplier:  USA Scientific Inc 40112321
Encompass
Eppendorf Tube 5.0 mL. Conical bottom centrifuge tube with screw cap. str, 2 bags of 100 tubes (200)
Catalog No. 50-274-4771