Learn More
USA Scientific Inc Eppendorf Tubes BioBased, 5 mL
Catalog No. 502744761
Encompass
Eppendorf Tubes BioBased, 5.0 mL, screw cap, str, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, colorless, 200 tubes (2 bags x 100 tubes)
Catalog No. 50-274-4761 Supplier USA Scientific Inc Supplier No. 40322518
Eppendorf Tubes BioBased, 5.0 mL, screw cap, str, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, colorless, 200 tubes (2 bags x 100 tubes)