Learn More
Medchemexpress LLC EPQpYEEIPIYL | 147612-86-0 | 99.5% | 1473.52 | 10 MG
SDP
Catalog No. 5000398582
Encompass_Preferred
EPQpYEEIPIYL is a phosphopeptide that functions as a Src homology 2 (SH2) domain ligand. It activates Src family members such as Lck, Hck, and Fyn by binding to their SH2 domains.
  • Functions as a phosphopeptide and a Src homology 2 (SH2) domain ligand.
  • Activates Src family members such as Lck, Hck, and Fyn.
  • Exhibits high-affinity binding to GST fusion proteins of the Lck SH2 domain and the Src SH2 domain.
  • For research use only.
Catalog No. 50-003-98582 Supplier Medchemexpress LLC Supplier No. HYP327910MG
EPQpYEEIPIYL is a phosphopeptide that functions as a Src homology 2 (SH2) domain ligand. It activates Src family members such as Lck, Hck, and Fyn by binding to their SH2 domains.
  • Functions as a phosphopeptide and a Src homology 2 (SH2) domain ligand.
  • Activates Src family members such as Lck, Hck, and Fyn.
  • Exhibits high-affinity binding to GST fusion proteins of the Lck SH2 domain and the Src SH2 domain.
  • For research use only.