Learn More
Medchemexpress LLC EPQpYEEIPIYL | 147612-86-0 | 99.5% | 1473.52 | 5 MG
SDP
Catalog No. 5000398350
Encompass_Preferred
EPQpYEEIPIYL, a phosphopeptide, is a Src homology 2 (SH2) domain ligand. It activates Src family members (e.g. Lck, Hck, Fyn) by binding to SH2 domains.
  • Phosphopeptide and Src homology 2 (SH2) domain ligand
  • Activates Src family members (e.g., Lck, Hck, Fyn)
  • Binds to SH2 domains
  • Exhibits highest affinity binding to GST fusion proteins of Lck SH2 domain and Src SH2 domain
  • Target: Src
  • Pathway: Protein Tyrosine Kinase/RTK
Catalog No. 50-003-98350 Supplier Medchemexpress LLC Supplier No. HYP32795MG
EPQpYEEIPIYL, a phosphopeptide, is a Src homology 2 (SH2) domain ligand. It activates Src family members (e.g. Lck, Hck, Fyn) by binding to SH2 domains.
  • Phosphopeptide and Src homology 2 (SH2) domain ligand
  • Activates Src family members (e.g., Lck, Hck, Fyn)
  • Binds to SH2 domains
  • Exhibits highest affinity binding to GST fusion proteins of Lck SH2 domain and Src SH2 domain
  • Target: Src
  • Pathway: Protein Tyrosine Kinase/RTK