Learn More
Norell Inc 4mm STANDARD EPR SAMPLE TUBE (LENGTH: 250mm), 5PK
SDP
Catalog No. NC9384526
Encompass_Preferred
4mm STANDARD EPR SAMPLE TUBE (LENGTH: 250mm), 5PK
Catalog No. NC9384526 Supplier Norell Inc Supplier No. S4EPR250S
4mm STANDARD EPR SAMPLE TUBE (LENGTH: 250mm), 5PK