Learn More
Medchemexpress LLC Eprazinone dihydrochloride | 10402-53-6 | 99.05% | 50 MG
SDP
Catalog No. 5000409775
Encompass_Preferred
Eprazinone dihydrochloride is a mucolytic, secretolytic, antitussic, and bronchial antispasmodic agent. It functions as a neurokinin 1 receptor (NK1R) ligand and shows potential for treating chronic bronchitis by improving pulmonary function and arterial partial pressure of oxygen. It is for research use only.
  • Functions as a neurokinin 1 receptor (NK1R) ligand
  • Potential for treating chronic bronchitis
  • Improves pulmonary function and arterial partial pressure of oxygen
  • Specifically displaces binding to the NK1R
  • May contribute to mucolytic activity through NK1R blockade
  • Increases total and individual phospholipid levels at higher doses in animal studies
  • Decreases total neutral lipids at higher doses in animal studies
  • Produces a dose-dependent decrease in short-circuit current (Isc) in airway epithelial studies
Catalog No. 50-004-09775 Supplier Medchemexpress LLC Supplier No. HYB2078A50MG
Eprazinone dihydrochloride is a mucolytic, secretolytic, antitussic, and bronchial antispasmodic agent. It functions as a neurokinin 1 receptor (NK1R) ligand and shows potential for treating chronic bronchitis by improving pulmonary function and arterial partial pressure of oxygen. It is for research use only.
  • Functions as a neurokinin 1 receptor (NK1R) ligand
  • Potential for treating chronic bronchitis
  • Improves pulmonary function and arterial partial pressure of oxygen
  • Specifically displaces binding to the NK1R
  • May contribute to mucolytic activity through NK1R blockade
  • Increases total and individual phospholipid levels at higher doses in animal studies
  • Decreases total neutral lipids at higher doses in animal studies
  • Produces a dose-dependent decrease in short-circuit current (Isc) in airway epithelial studies