Learn More
Medchemexpress LLC Eprazinone dihydrochloride | 10402-53-6 | 99.1% | 100 MG
SDP
Catalog No. 5000410000
Encompass_Preferred
Eprazinone dihydrochloride is an agent possessing mucolytic, secretolytic, antitussive, and bronchial antispasmodic properties. It acts as a neurokinin 1 receptor (NK1R) ligand. This compound shows potential for treating chronic bronchitis by improving pulmonary function and arterial partial pressure of oxygen.
  • Functions as a neurokinin 1 receptor (NK1R) ligand.
  • Exhibits mucolytic properties.
  • Possesses secretolytic characteristics.
  • Acts as an antitussive agent.
  • Displays bronchial antispasmodic effects.
  • Potential to improve pulmonary function.
  • Potential to improve arterial partial pressure of oxygen.
Catalog No. 50-004-10000 Supplier Medchemexpress LLC Supplier No. HYB2078A100MG
Eprazinone dihydrochloride is an agent possessing mucolytic, secretolytic, antitussive, and bronchial antispasmodic properties. It acts as a neurokinin 1 receptor (NK1R) ligand. This compound shows potential for treating chronic bronchitis by improving pulmonary function and arterial partial pressure of oxygen.
  • Functions as a neurokinin 1 receptor (NK1R) ligand.
  • Exhibits mucolytic properties.
  • Possesses secretolytic characteristics.
  • Acts as an antitussive agent.
  • Displays bronchial antispasmodic effects.
  • Potential to improve pulmonary function.
  • Potential to improve arterial partial pressure of oxygen.