Learn More
Epredia™ Epredia™ Nova Hopper
Catalog No. 22028547 Shop All Epredia Products
Change view
Click to view available options
Product Type:
NOVA Additional Hopper
Catalog No. Product Type
22-028-547 NOVA Additional Hopper
1 options
Catalog No. 22-028-547 Supplier Epredia™ Supplier No. 9001497
Add to Cart
Add to Cart
Laser cassette printer with 80-cassette capacity
  • Designed for use with the Epredia™ Epredia™ Nova laser cassette printer
  • Holds up to 80 additional cassettes
  • Allows for for easy exchange to a different color or style of cassettes
Specifications
Product Type NOVA Additional Hopper