Learn More
Medchemexpress LLC HY-19980 25mg , Eprenetapopt CAS:5291-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1998025MG
Encompass_Preferred
HY-19980 25mg Eprenetapopt CAS: 5291-32-7 Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. Eprenetapopt triggers apoptosis in tumor cells. Eprenetapopt also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9562