Learn More
Medchemexpress LLC Eprinomectin | 123997-26-2 | 98.02% | 914.13 | 250 MG
Supplier: Medchemexpress LLC 250MGHY12643
Eprinomectin (MK-397) is an avermectin type that acts as a broad-spectrum fungicide with insecticidal, insecticidal, and acaricidal properties. It also induces apoptosis and autophagy in prostate cancer cells, demonstrating antitumor activity.
- Broad-spectrum fungicide
- Insecticidal, insecticidal and acaricidal activities
- Induces apoptosis and autophagy in prostate cancer cells
- Antitumor activity