Learn More
Apexbio Technology LLC EPRINOMECTIN-250MG
SDP
Catalog No. 5000569091
Encompass_Preferred
Eprinomectin (CAS No 123997-26-2) is an avermectin derivative primarily studied in the field of parasitology and veterinary medicine Originating from the fermentation of Streptomyces avermitilis it functions by binding to glutamate-gated chloride channels in nerve and muscle cells of parasites leading to paralysis and death Eprinomectin exhibits notable in vitro activity in the nanomolar to low micromolar range making it effective against a wide array of nematodes and ectoparasites Typical applications involve cell models and animal studies to explore its anthelmintic properties as well as in drug screening for antiparasitic agents Experimental concentrations typically depend on the specific research design allowing for flexibility in dosage determination
Catalog No. 50-005-69091 Supplier Apexbio Technology LLC Supplier No. BA1268250
Add to Cart
Add to Cart
Eprinomectin (CAS No 123997-26-2) is an avermectin derivative primarily studied in the field of parasitology and veterinary medicine Originating from the fermentation of Streptomyces avermitilis it functions by binding to glutamate-gated chloride channels in nerve and muscle cells of parasites leading to paralysis and death Eprinomectin exhibits notable in vitro activity in the nanomolar to low micromolar range making it effective against a wide array of nematodes and ectoparasites Typical applications involve cell models and animal studies to explore its anthelmintic properties as well as in drug screening for antiparasitic agents Experimental concentrations typically depend on the specific research design allowing for flexibility in dosage determination