Learn More
Medchemexpress LLC Eprodisate (70% in water) | 21668-77-9 | 98.0% | 1 ML
SDP
Catalog No. 5000338436
Encompass_Preferred
Eprodisate (70% in water) is an amyloid inhibitor that interferes with the interaction between amyloidogenic proteins and glycosaminoglycans. It is intended for research use only.
  • Inhibits aggregation of amyloid fibrils
  • Inhibits deposition of fibrils in tissues
  • Slows progression of renal disease associated with AA amyloidosis
  • May be useful for other types of amyloidosis
Catalog No. 50-003-38436 Supplier Medchemexpress LLC Supplier No. HY12884910MM/1ML
Eprodisate (70% in water) is an amyloid inhibitor that interferes with the interaction between amyloidogenic proteins and glycosaminoglycans. It is intended for research use only.
  • Inhibits aggregation of amyloid fibrils
  • Inhibits deposition of fibrils in tissues
  • Slows progression of renal disease associated with AA amyloidosis
  • May be useful for other types of amyloidosis