Learn More
Selleck Chemical LLC Eprodisate disodium S4567-100mg
SDP
Catalog No. 503110444
Encompass_Preferred
Eprodisate Disodium (NC-503) is the orally available disodium salt form of Eprodisate a small-molecule orally bioavailable inhibitor of AA-amyloid fibrillogenesis
Catalog No. 50-311-0444 Supplier Selleck Chemical LLC Supplier No. S4567100MG
Eprodisate Disodium (NC-503) is the orally available disodium salt form of Eprodisate a small-molecule orally bioavailable inhibitor of AA-amyloid fibrillogenesis