Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eprodisate disodium | 36589-58-9 | 98.0% | 1 ML
SDP

Catalog No. 5000413102
Encompass_Preferred

Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate, a negatively charged sulfonated inhibitor of fibrillogenesis, that can be used in the treatment of amyloid A (AA) amyloidosis.

  • Appearance: solid
  • Color: white to off-white
  • Solubility (in vitro): H2O: 50 mg/mL (201.46 mM; need ultrasonic)
  • Solubility (in vitro): DMSO: 7.69 mg/mL (30.98 mM; need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
  • Orally available disodium salt form of Eprodisate
  • Negatively charged sulfonated inhibitor of fibrillogenesis
  • Can be used in the treatment of amyloid A (AA) amyloidosis

Catalog No. 50-004-13102 Supplier Medchemexpress LLC Supplier No. HYB114110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate, a negatively charged sulfonated inhibitor of fibrillogenesis, that can be used in the treatment of amyloid A (AA) amyloidosis.

  • Appearance: solid
  • Color: white to off-white
  • Solubility (in vitro): H2O: 50 mg/mL (201.46 mM; need ultrasonic)
  • Solubility (in vitro): DMSO: 7.69 mg/mL (30.98 mM; need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
  • Orally available disodium salt form of Eprodisate
  • Negatively charged sulfonated inhibitor of fibrillogenesis
  • Can be used in the treatment of amyloid A (AA) amyloidosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.