Learn More
Medchemexpress LLC Eprodisate disodium | 36589-58-9 | 98.0% | 500 MG
SDP
Catalog No. 5000413348
Encompass_Preferred
Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate. It is a negatively charged sulfonated inhibitor of fibrillogenesis that can be used in the treatment of amyloid A (AA) amyloidosis.
  • Orally available disodium salt form of Eprodisate.
  • Negatively charged sulfonated inhibitor of fibrillogenesis.
  • Used in treatment of amyloid A (AA) amyloidosis.
  • Targets Amyloid-β.
  • Involves Neuronal Signaling pathway.
Catalog No. 50-004-13348 Supplier Medchemexpress LLC Supplier No. HYB1141500MG
Eprodisate disodium (NC-503) is the orally available disodium salt form of Eprodisate. It is a negatively charged sulfonated inhibitor of fibrillogenesis that can be used in the treatment of amyloid A (AA) amyloidosis.
  • Orally available disodium salt form of Eprodisate.
  • Negatively charged sulfonated inhibitor of fibrillogenesis.
  • Used in treatment of amyloid A (AA) amyloidosis.
  • Targets Amyloid-β.
  • Involves Neuronal Signaling pathway.