Learn More
Medchemexpress LLC Eprosartan | 133040-01-4 | MFCD00897872 | 424.5 g/mol | C23H24N2O4S | 10 MG
SDP
Catalog No. 5000227881
Encompass_Preferred
Eprosartan is a selective, competitive, orally active angiotensin II receptor antagonist used in research on antihypertensive mechanisms and receptor pharmacology. The compound is typically supplied as a white to off-white crystalline powder for analytical and in vivo studies.
  • Selective angiotensin II receptor antagonist with documented receptor binding activity.
  • Orally active compound suitable for in vivo pharmacology studies.
  • Supplied as a crystalline free base appropriate for analytical and research use.
  • Well-characterized in public chemical databases for unambiguous identification.
Catalog No. 50-002-27881 Supplier Medchemexpress LLC Supplier No. HY11774310MG
Eprosartan is a selective, competitive, orally active angiotensin II receptor antagonist used in research on antihypertensive mechanisms and receptor pharmacology. The compound is typically supplied as a white to off-white crystalline powder for analytical and in vivo studies.
  • Selective angiotensin II receptor antagonist with documented receptor binding activity.
  • Orally active compound suitable for in vivo pharmacology studies.
  • Supplied as a crystalline free base appropriate for analytical and research use.
  • Well-characterized in public chemical databases for unambiguous identification.