Learn More
TARGETMOL CHEMICALS INC Eprosartan Mesylate 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2531100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Eprosartan Mesylate (SKF-108566J) is a competitive and reversible Angiotensin II Receptor Blocker. Purity 100%
Catalog No. 50-287-8230