Learn More
Apexbio Technology LLC Eprosartan Mesylate 144143-96-4 50mg
SDP
Catalog No. 5000091766
Encompass_Preferred
Eprosartan Mesylate is a small-molecule antagonist targeting the angiotensin II AT1 receptor It is designed to selectively inhibit the AT1 receptor subtype thereby modulating vasoconstriction and aldosterone secretion mechanisms in cardiovascular systems Eprosartan Mesylate exerts its biological activity primarily through competitive binding to AT1 receptors interfering with angiotensin II action In studies using rat and human adrenal cortical membranes Eprosartan Mesylate demonstrates inhibitory activity with IC50 values of approximately 9 2 nM and 3 9 nM respectively Based on these pharmacological properties Eprosartan Mesylate holds research potential in angiotensin II-mediated signal transduction pathways hypertension models and related cardiovascular diseases
Catalog No. 50-000-91766 Supplier Apexbio Technology LLC Supplier No. B174650
Eprosartan Mesylate is a small-molecule antagonist targeting the angiotensin II AT1 receptor It is designed to selectively inhibit the AT1 receptor subtype thereby modulating vasoconstriction and aldosterone secretion mechanisms in cardiovascular systems Eprosartan Mesylate exerts its biological activity primarily through competitive binding to AT1 receptors interfering with angiotensin II action In studies using rat and human adrenal cortical membranes Eprosartan Mesylate demonstrates inhibitory activity with IC50 values of approximately 9 2 nM and 3 9 nM respectively Based on these pharmacological properties Eprosartan Mesylate holds research potential in angiotensin II-mediated signal transduction pathways hypertension models and related cardiovascular diseases