Learn More
Medchemexpress LLC Eprosartan mesylate | 144143-96-4 | MFCD08141807 | 100.0% | 520.62 g/mol | C24H28N2O7S2 | 10 MG
SDP
Catalog No. 5000288675
Encompass_Preferred
Eprosartan mesylate is a selective, competitive, orally active angiotensin II receptor antagonist supplied for research use in pharmacology and biochemical studies. It is provided as a high-purity solid and as solution formulations for in vitro assays, and is supported by technical documentation (datasheet, COA, SDS) for laboratory handling and experimental planning.
  • Selective angiotensin II receptor antagonist suitable for receptor binding and functional assays.
  • High purity appropriate for research applications (listed at 99.95%).
  • Available as a solid and as a 10 mM solution in DMSO for in vitro use.
  • Supported by datasheet, certificate of analysis, and safety data sheet.
  • Stable when stored sealed and protected from moisture; specific solvent storage conditions available.
Catalog No. 50-002-88675 Supplier Medchemexpress LLC Supplier No. HY15834A10MG
Eprosartan mesylate is a selective, competitive, orally active angiotensin II receptor antagonist supplied for research use in pharmacology and biochemical studies. It is provided as a high-purity solid and as solution formulations for in vitro assays, and is supported by technical documentation (datasheet, COA, SDS) for laboratory handling and experimental planning.
  • Selective angiotensin II receptor antagonist suitable for receptor binding and functional assays.
  • High purity appropriate for research applications (listed at 99.95%).
  • Available as a solid and as a 10 mM solution in DMSO for in vitro use.
  • Supported by datasheet, certificate of analysis, and safety data sheet.
  • Stable when stored sealed and protected from moisture; specific solvent storage conditions available.