Learn More
Cayman Chemical Eprosartan (mesylate), Purity: ≥98%, CAS: 144143-96-4, 100 mg
SDP
Catalog No. 502951779
Encompass_Preferred
A competitive AT1 receptor antagonist (IC50s 1.4 – 3.9 nM and an elimination half-life of 5 to 7 hours) that blocks angiotensin II receptors on both sympathetic nerve terminals and blood vessels
Catalog No. 50-295-1779 Supplier Cayman Chemical Supplier No. 11578100 MG
A competitive AT1 receptor antagonist (IC50s 1.4 – 3.9 nM and an elimination half-life of 5 to 7 hours) that blocks angiotensin II receptors on both sympathetic nerve terminals and blood vessels