Learn More
Cayman Chemical Eprosartnmesylate 10mg
SDP
Catalog No. 502953313
Encompass_Preferred
A competitive AT1 receptor antagonist (IC50s 1.4 – 3.9 nM and an elimination half-life of 5 to 7 hours) that blocks angiotensin II receptors on both sympathetic nerve terminals and blood vessels
Catalog No. 50-295-3313 Supplier Cayman Chemical Supplier No. 1157810 MG
A competitive AT1 receptor antagonist (IC50s 1.4 – 3.9 nM and an elimination half-life of 5 to 7 hours) that blocks angiotensin II receptors on both sympathetic nerve terminals and blood vessels