Learn More
Aviva Systems Biology Corp EPRS Antibody - middle region (ARP46109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPRS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPRS. Alternate names: DKFZp313B047, EARS, PARS, PIG32, QARS, QPRS, GLUPRORSApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59229