Learn More
SOMNI Scientific™ EPS-3 Exposure Prevention System 3 Station Flowmeter Controlled Attenuation
SDP
Catalog No. 04777168 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-168 Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-168 Supplier SOMNI Scientific™ Supplier No. WG15002
Add to Cart
Add to Cart
Can be used to attenuate in-house active vacuum to safely remove anesthetic waste gas from multiple devices simultaneously
  • Requires in-house active vacuum system
  • Attenuates the vacuum from multiple devices for effective waste gas removal
  • Works with active style accessories
  • Prevents unnecessary exposure
Specifications
For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15003, MW-17005
Material Powder-coated Aluminum
Length (Metric) 30 cm
Width (Metric) 20 cm
Includes Evacuation lines and outlet tubing with adapters
For Veterinary Use Only