Learn More
Aviva Systems Biology Corp EPS8 Antibody - middle region (ARP54726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPS8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPS8. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61292