Learn More
Medchemexpress LLC Epsilon-caprolactone | 502-44-3 | MFCD00003267 | 100.0% | 114.14 g/mol | C6H10O2 | 1 KG
SDP
Catalog No. 5000303885
Encompass_Preferred
Epsilon-caprolactone is a cyclic ester (lactone) monomer widely used in organic synthesis and polymer chemistry, notably for producing polycaprolactone via ring-opening polymerization. It is a colorless to light yellow liquid (C6H10O2, MW 114.14) with CAS 502-44-3 and is supplied at high purity for research and materials synthesis.
  • Used as a monomer for ring-opening polymerization.
  • Enables synthesis of biodegradable polycaprolactone.
  • High purity suitable for research applications.
  • Liquid at room temperature for easy handling.
  • Reacts readily in nucleophilic ring-opening reactions.
Catalog No. 50-003-03885 Supplier Medchemexpress LLC Supplier No. HYW0158911KG
Epsilon-caprolactone is a cyclic ester (lactone) monomer widely used in organic synthesis and polymer chemistry, notably for producing polycaprolactone via ring-opening polymerization. It is a colorless to light yellow liquid (C6H10O2, MW 114.14) with CAS 502-44-3 and is supplied at high purity for research and materials synthesis.
  • Used as a monomer for ring-opening polymerization.
  • Enables synthesis of biodegradable polycaprolactone.
  • High purity suitable for research applications.
  • Liquid at room temperature for easy handling.
  • Reacts readily in nucleophilic ring-opening reactions.