Learn More
Medchemexpress LLC Epsilon-caprolactone | 502-44-3 | MFCD00003267 | 100.0% | 114.14 g/mol | C6H10O2 | 100 G
SDP
Catalog No. 5000299624
Encompass_Preferred
Epsilon-caprolactone (C6H10O2; MW 114.14 g/mol; CAS 502-44-3) is a six-membered lactone used as a monomer and synthetic building block in organic and polymer chemistry. It is a colorless liquid with high assay suitable for research and synthesis, commonly employed in ring-opening polymerization to produce biodegradable polyesters and copolymers.
  • High purity suitable for research-grade synthesis.
  • Liquid monomer readily polymerized via ring-opening polymerization.
  • Versatile building block for producing biodegradable polyesters and copolymers.
  • Compatible with standard organic solvents and common reaction conditions.
  • Available with certificate of analysis and safety data sheet for reproducibility and safety.
Catalog No. 50-002-99624 Supplier Medchemexpress LLC Supplier No. HYW015891100G
Epsilon-caprolactone (C6H10O2; MW 114.14 g/mol; CAS 502-44-3) is a six-membered lactone used as a monomer and synthetic building block in organic and polymer chemistry. It is a colorless liquid with high assay suitable for research and synthesis, commonly employed in ring-opening polymerization to produce biodegradable polyesters and copolymers.
  • High purity suitable for research-grade synthesis.
  • Liquid monomer readily polymerized via ring-opening polymerization.
  • Versatile building block for producing biodegradable polyesters and copolymers.
  • Compatible with standard organic solvents and common reaction conditions.
  • Available with certificate of analysis and safety data sheet for reproducibility and safety.