Learn More
Medchemexpress LLC Epsilon-caprolactone | 502-44-3 | MFCD00003267 | 100.0% | 114.14 g/mol | C6H10O2 | 500 G
SDP
Catalog No. 5000299622
Encompass_Preferred
ε-Caprolactone is a cyclic ester (lactone) monomer used in ring-opening polymerization to produce polycaprolactone and related polymers. It is used as a building block in organic synthesis and materials research, providing a low-viscosity liquid reagent for polymer chemistry and formulation studies.
  • High purity (99.98%).
  • Chemical formula C6H10O2, molecular weight 114.14 g/mol.
  • Liquid appearance, colorless to light yellow, density ≈1.16 g/cm³.
  • Suitable for ring-opening polymerization to produce polycaprolactone.
  • Commonly used in polymer synthesis, materials research, and organic synthesis.
Catalog No. 50-002-99622 Supplier Medchemexpress LLC Supplier No. HYW015891500G
ε-Caprolactone is a cyclic ester (lactone) monomer used in ring-opening polymerization to produce polycaprolactone and related polymers. It is used as a building block in organic synthesis and materials research, providing a low-viscosity liquid reagent for polymer chemistry and formulation studies.
  • High purity (99.98%).
  • Chemical formula C6H10O2, molecular weight 114.14 g/mol.
  • Liquid appearance, colorless to light yellow, density ≈1.16 g/cm³.
  • Suitable for ring-opening polymerization to produce polycaprolactone.
  • Commonly used in polymer synthesis, materials research, and organic synthesis.