Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epsilon-(gamma-glutamyl)-lysine TFA | 00-00-0 | MFCD00055787 | 99.9% | 503.35 g·mol⁻¹ | C15H23F6N3O9 | 5 MG
SDP

Catalog No. 5000262477
Encompass_Preferred

Epsilon-(gamma-glutamyl)-lysine TFA is the trifluoroacetate salt of epsilon-(gamma-glutamyl)-lysine supplied as a white to off-white solid for research use. It is intended for in vitro and in vivo formulation studies where the TFA salt form is required and includes stability and solubility guidance for experimental workflows.

  • High purity (99.9%).
  • TFA salt form for improved handling in formulation studies.
  • Solid, white to off-white appearance.
  • Molecular weight 503.35 g·mol⁻¹; formula C15H23F6N3O9.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Recommended storage: powder -80°C (2 years) or -20°C (1 year) when sealed.

Catalog No. 50-002-62477 Supplier Medchemexpress LLC Supplier No. HY113089A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epsilon-(gamma-glutamyl)-lysine TFA is the trifluoroacetate salt of epsilon-(gamma-glutamyl)-lysine supplied as a white to off-white solid for research use. It is intended for in vitro and in vivo formulation studies where the TFA salt form is required and includes stability and solubility guidance for experimental workflows.

  • High purity (99.9%).
  • TFA salt form for improved handling in formulation studies.
  • Solid, white to off-white appearance.
  • Molecular weight 503.35 g·mol⁻¹; formula C15H23F6N3O9.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Recommended storage: powder -80°C (2 years) or -20°C (1 year) when sealed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.