Learn More
Medchemexpress LLC HY-111634 10mg Medchemexpress, Epsilon-momfluorothrin CAS:1065124-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11163410MG
Encompass_Preferred
Medchemexpress, HY-111634 10mg Epsilon-momfluorothrin CAS:1065124-65-3 Epsilon-momfluorothrin is a type I synthetic pyrethroid insecticide, activates constitutive androstane receptor (CAR), and induces hepatocellular tumors in rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8929