Learn More
TARGETMOL CHEMICALS INC EPSILON-MOMFLUOROTHRIN 5MG
SDP
Catalog No. 502581332
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Epsilon-momfluorothrin (Momfluorothrin (Z1R3R)-) a type I synthetic pyrethroid insecticide exerts its insecticidal properties and triggers activation of the constitutive androstane receptor (CAR). Notably its administration in rats has been observed to induce the development of hepatocellular tumors. purity: 99%
Catalog No. 50-258-1332 Supplier TARGETMOL CHEMICALS INC Supplier No. T136835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Epsilon-momfluorothrin (Momfluorothrin (Z1R3R)-) a type I synthetic pyrethroid insecticide exerts its insecticidal properties and triggers activation of the constitutive androstane receptor (CAR). Notably its administration in rats has been observed to induce the development of hepatocellular tumors. purity: 99%