Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epsilon-Thiocaprolactam 98.0+%, TCI America™
SDP

Catalog No. T129710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T129710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T129710G Supplier TCI America Supplier No. T129710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7203-96-5
Molecular Formula C6H11NS
Molecular Weight (g/mol) 129.221
MDL Number MFCD00006937
InChI Key APLHDUWNMGJBFD-UHFFFAOYSA-N
Synonym Hexahydro-2-azepinethione, 2-Thioxohexamethyleneimine
PubChem CID 3003736
IUPAC Name azepane-2-thione
SMILES C1CCC(=S)NCC1

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 171°C
Quantity 10 g
Formula Weight 129.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material epsilon-Thiocaprolactam
RTECSNumber CM3370000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.