Learn More
Selleck Chemical LLC Epsilon-V1-2 P1160-5MG
SDP
Catalog No. 5000040859
Encompass_Preferred
Epsilon-V1-2 ( -V1-2) a PKC -derived peptide is a selective PKC inhibitor which can inhibit the translocationof PKC but not - - and -PKC
Catalog No. 50-000-40859 Supplier Selleck Chemical LLC Supplier No. P11605MG
Epsilon-V1-2 ( -V1-2) a PKC -derived peptide is a selective PKC inhibitor which can inhibit the translocationof PKC but not - - and -PKC