Learn More
CORMARK LLC EPSON C7500 MATTE BLACK INKJET
Catalog No. NC3361867
Encompass
  • High-performance pigment-based ink solution
  • Utilizes DURABrite Ultra pigment ink technology
  • Water resistant, chemical resistant, smudge resistant, and fade resistant
  • Compatible with Epson ColorWorks TM-C7500 (matte) label printer
  • Ideal for GHS-compliant chemical labeling applications
  • Delivers sharp, crisp, clear prints
  • Fast-drying for efficient labeling
  • Designed to minimize waste and reduce user interventions
  • High-capacity cartridge volume of 340 ml
  • BS-5609 certified when used with appropriate media
Catalog No. NC3361867 Supplier CORMARK LLC Supplier No. CWC7500M
Add to Cart
Add to Cart
Request a Quote
  • High-performance pigment-based ink solution
  • Utilizes DURABrite Ultra pigment ink technology
  • Water resistant, chemical resistant, smudge resistant, and fade resistant
  • Compatible with Epson ColorWorks TM-C7500 (matte) label printer
  • Ideal for GHS-compliant chemical labeling applications
  • Delivers sharp, crisp, clear prints
  • Fast-drying for efficient labeling
  • Designed to minimize waste and reduce user interventions
  • High-capacity cartridge volume of 340 ml
  • BS-5609 certified when used with appropriate media