Learn More
ACROBiosystems ACROBiosystems Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 / EBV GP350 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems GP0E52H6100UG
Encompass_Preferred
Recombinant Protein;100UG;Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 Protein, His Tag (GP0-E52H6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 470 (Accession # P03200-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.3 kDa. The protein migrates as 80-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6945