Learn More
Medchemexpress LLC Epstein-Barr virus (EBV) lytic cycle inducer-1 Dp44mT (compound C7) | 394668-43-0 | 98.3% | 318.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495775MG
Encompass_Preferred
This Epstein-Barr virus (EBV) lytic cycle inducer is an iron-chelator-like compound that reactivates the EBV lytic cycle by activating the ERK1/2-autophagy axis in epithelial cancers. It can cooperate with HDAC inhibitors to induce the EBV lytic cycle.
  • Induces lytic cycle in a cell line-dependent manner
  • Activates the ERK1/2-autophagy axis
  • Higher toxicity in EBV-positive cell lines
Catalog No. 50-003-59489