Learn More
Bioworld EPSTEIN-BARR VIRUS (HHV-4)
Supplier:  Bioworld 220600631
Encompass_Preferred
EPSTEIN-BARR VIRUS (HHV-4) EBNA1, (RECOMBINANT): Recombinant EBV (HHV-4) Nuclear Protein EBNA1 Source: E.Coli Background: The Epstein-Barr virus (EBV), also called Human herpes virus 4 (HHV-4), is a virus of the herpes family (which in...
Catalog No. 50-253-160