Learn More
Bioworld Epstein-Barr Virus (HHV-4) p18 (Recombinant), 100 μg
Supplier:  Bioworld 220600641
Encompass_Preferred
Recombinant Epstein-Barr Virus Capsid Antigen p18 Source: E.Coli Description: Recombinant Epstein-Barr Virus protein contains the EBV(HHV-4) p18 regions,amino acids:(1-119). Purification method: Recombinant EBV is purified by proprietary chromatographic technique. Purity: Recombinant EBV is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein concentration: 1mg/ml in 50mM Tris-HCl, pH 8.0, 60 mM NaCland 50% glycerol. Storage: Recombinant EBV is shipped at ambient temperature. Upon arrival, Store at -20°C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of EBV-infected individuals. Application: Antigen may be used in: 1.ELISA 2.Western blots 3.Detection of HHV-4 (EBV) with minimal specificity problems. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals
Catalog No. 50-253-161
 Restricted Item