Learn More
StatLab Epstein Barr Virus IHC Control Slides
SDP
Catalog No. 22024920
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22-024-920 25 Pk.
1 options
Catalog No. 22-024-920 Supplier StatLab Supplier No. CSE0125P
Add to Cart
Add to Cart
Use epstein barr virus control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Sterility Non-sterile
Quantity 25 Pk.