Learn More
Alpco Ltd Epstein Barr Virus Nuclear Antigen IgG ELISA
SDP
Supplier:  Alpco Ltd 20ENGHUE01
Encompass_Preferred
The Epstein-Barr Virus (EBNA-1) IgG Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgG-class antibodies to Epstein-Barr virus nuclear antigen type 1 (EBV-EBNA-1) in human serum or plasma (EDTA, heparin, citrate). Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-091