Learn More
Crescent Chemical Co Inc EPTC, 759-94-4, MFCD00053780, 1mL
SDP
Supplier:  Crescent Chemical Co Inc CT7507A.10
Encompass_Preferred
Molecular Formula C9H19NOS, Molecular Weight 189.321, 1000ppm, Single Component Calibration Standard in Acetone
Catalog No. 50-709-48