Learn More
Medchemexpress LLC HY-19946 5mg Medchemexpress, Eptapirone CAS:179756-58-2 Purity:>98%
SDP
Catalog No. 502082199
Encompass_Preferred
Medchemexpress, HY-19946 5mg Eptapirone CAS:179756-58-2 Eptapirone (F11440) is a potent, selective, high efficacy 5-HT1A receptor agonist with marked anxiolytic and antidepressant potential. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2199 Supplier Medchemexpress LLC Supplier No. HY199465MG
Medchemexpress, HY-19946 5mg Eptapirone CAS:179756-58-2 Eptapirone (F11440) is a potent, selective, high efficacy 5-HT1A receptor agonist with marked anxiolytic and antidepressant potential. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.