Learn More
Bachem Eptifibatide Acetate
SDP
Supplier:  Bachem 4063191.0001
Encompass_Preferred
Eptifibatide Acetate, 3-Mercaptopropionyl-Homoarg-Gly-Asp-Trp-Pro-Cys-NH2, 188627-80-7, C35H49N11O9S2, Mr 831.97, 1mg. Eptifibatide, a RGD mimetic, is a glycoprotein (GP) IIb/IIIa receptor antagonist. Clinically, the peptide is used as an antiplatelet drug in combination with heparin and aspirin in the management of unstable angina and in patients undergoing coronary angioplasty and stenting procedures. CAS Number (eptifibatide acetate): 148031-34-9.
Catalog No. 50-383-299